Pure G3 Sterile Latex Gloves
Actual product may vary
Pure G3 Sterile Latex Gloves
Sterile, hand-specific latex gloves are suitable for double-donning and provide sensitive protection. Kimtech™ Pure G3 Sterile Latex Gloves recommended for Class 10/ISO 4 or higher cleanrooms.
Have Questions?
Catalog NumberSize
KLC568467.5
Catalog number KLC56846
Price (SGD)
420.00
Special offer
Ends: 31-Dec-2026
578.00
Save 158.00 (27%)
Case
Size:
7.5
Price (SGD)
420.00
Special offer
Ends: 31-Dec-2026
578.00
Save 158.00 (27%)
Case
Sterile, hand-specific latex gloves are suitable for double-donning and provide sensitive protection. Recommended for Class 10/ISO 4 or higher cleanrooms.
  • Fully sterilized latex material provides high levels of contamination protection
  • Hand-specific, disposable and a natural color
  • Textured palm and fingertips enhance grip and tactile sensitivity
  • Beaded cuffs add strength to the gloves reducing the risk of tearing
  • Easy donning and doffing
  • Made from latex sterilized with gamma radiation and containing no powder
  • Suitable for use in Class 10/ISO 4 sterile cleanrooms
Specifications
Certifications/ComplianceASTM D3577, ASTM D5151, ASTM D412, ASTM D573, ASTM D3767, ASTM D6319, IEST-RP-CC005, ASTM D5712
Certifications/Compliance (Europe)EN ISO 374-1:2016, EN 374-4:2003, EN ISO 374-5:2016
Cleanroom ClassClass 10 (ISO 4)
Accelerator FreeYes
AQL1.5
ColorNatural
DisposableSingle-use
Exterior FinishFully Textured
Finger Thickness (English)8.7 mil
Finger Thickness (Metric)0.22 mm
HandSpecific
Laboratory/Cleanroom UseControlled Environments
Latex FreeNo
MaterialLatex
Palm Thickness (English)7.9 mil
Palm Thickness (Metric)0.20 mm
PowderedPowder Free
Product TypeCleanroom Gloves
Size7.5
SterilitySterile
Thickness (English)8 mil
Thickness (Metric)0.20 mm
Model56846
SeriesKimtech Pure G3
Tensile Strength24 MPa
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCase