1x1 image pixel for data collection
Kimtech™ Purple Nitrile-Xtra™ Gloves
Kimtech™ Purple Nitrile-Xtra™ Gloves
Actual product may vary
Kimtech™ Purple Nitrile-Xtra™ Gloves
Kimtech™ Purple Nitrile-Xtra™ Gloves
Kimtech™ Purple Nitrile-Xtra™ Gloves
Kimtech™ Purple Nitrile-Xtra™ Gloves
Purple Nitrile Xtra™ Gloves are 12 in. in length with a beaded cuff to provide maximum protection for higher-risk procedures.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC55090X-Small
KLC50601Small
KLC50602Medium
KLC50603Large
KLC50604X-Large
Catalog number KLC55090
Price (SGD)
235.00
CS
Add to cart
Size:
X-Small
Price (SGD)
235.00
CS
Add to cart
Purple Nitrile Xtra™ Gloves are 12 in. in length with a beaded cuff to provide maximum protection for higher-risk procedures.
  • Textured fingertips offers dexterity and tactile sensitivity
  • Latex-free
  • Ambidextrous
  • 12 in. length
  • Beaded cuff prevent roll-down and allow for easy donning
  • Powder free
Specifications
Certifications/ComplianceASTM D6978-05 (non-sterile only), Meets USP <800> guidelines, Meets NFPA standard 1999-2018
Chemotherapeutic RatedYes
ApplicationsHealthcare, Laboratory
AQL0.65
ColorPurple
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)6 mil
Finger Thickness (Metric)0.15 mm
Latex FreeYes
MaterialNitrile
Palm Thickness (English)4.7 mil
Palm Thickness (Metric)0.11 mm
PowderedPowder Free
SizeX-Small
Thickness (English)6 mil
Thickness (Metric)0.15 mm
FDA 510 (k)Yes
Model55090
SeriesKimtech™
Tensile Strength21 MPa
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCS
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link