Tubing adaptor is a national pipe thread taper made up of stainless steel which is used to connect a female adaptor. It can be used to connect capillary tubes in HPLC, GC.
Have Questions?
Catalog Number
Packaging Quantity
KLC55082
1/Pk.
Catalog number KLC55082
Price (SGD)
26.45
Special offer
Ends: 16-Jul-2026
362.00
Save 335.55 (93%)
Case
Packaging Quantity:
1/Pk.
Price (SGD)
26.45
Special offer
Ends: 16-Jul-2026
362.00
Save 335.55 (93%)
Case
Tefzel is a registered trademark of E. I. du Pont de Nemours and Company