Kimtech™ Pure G3 EvT Nitrile Gloves
Kimtech™ Pure G3 EvT Nitrile Gloves
Actual product may vary
Kimtech™ Pure G3 EvT Nitrile Gloves
Non-sterile, nitrile gloves designed to have low particles and extractables and are cleaned to meet the needs of a ISO Class 3 or higher critical environment.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC38704X-Large
KLC38700X-Small
KLC38701Small
KLC38702Medium
KLC38703Large
Catalog number KLC38704
Price (SGD)
270.00
Special offer
414.00
Save 144.00 (35%)
CS
Add to cart
Size:
X-Large
Price (SGD)
270.00
Special offer
414.00
Save 144.00 (35%)
CS
Add to cart
  • Natural rubber latex-free reduces the risk of Type I allergic reactions
  • Recommended for ISO Class 3 and higher environments
  • Low particles and extractables
  • Cleaned to meet the needs of the critical cleanroom environment
  • Textured and tacky surface provides secure grip
  • AQL 1.5 for pinholes
  • Ambidextrous
  • Ideal for critical tasks in semiconductor, electronics and medical device manufacturing facilities
Specifications
Certifications/ComplianceASTM D5151
Cleanroom ClassClass 1 (ISO 3)
AQL1.5
ColorWhite
Exterior FinishFully Textured
Finger Thickness (English)5 mil
Finger Thickness (Metric)0.12 mm
HandAmbidextrous
Latex FreeYes
MaterialNitrile
SizeX-Large
SterilityNon-sterile
Thickness (English)5 mil
Thickness (Metric)0.12 mm
Model38704
SeriesKimtech™ Pure G3 EvT
Length (English)12 in.
Length (Metric)30.5 cm
Unit SizeCS