A5 Sterile Integrated Hood and Mask
Actual product may vary
Ansell
A5 Sterile Integrated Hood and Mask
Cleanroom sterile hood with a mask integrated into the hood.
Have Questions?
Catalog NumberSize
KLC36072Universal
Catalog number KLC36072
Price (SGD)
640.00
Special offer
Ends: 06-Sep-2026
777.00
Save 137.00 (18%)
Case
Size:
Universal
Price (SGD)
640.00
Special offer
Ends: 06-Sep-2026
777.00
Save 137.00 (18%)
Case
Cleanroom sterile hood with a mask integrated into the hood.
  • Average bacterial filtration efficiency (BFE) of 96%
  • Polypropylene breathable SMS fabric
  • Triple-bagged and vacuum packaged
  • Suitable for ISO Class 5 or higher cleanrooms
  • Clean-don ties for easy gowning and are folded inside out for aseptic donning
  • Protective hood-and-mask-combos are made without latex, silicone or the BHT preservative
  • Designed for single-use
  • CE certified, Category III
  • Each individually bagged mask and hood has a sterility indicator
  • Manufactured in an ISO-certified facility, clean-processes and individually packaged under cleanroom conditions
Specifications
Product TypeIntegrated Hood and Mask
SizeUniversal
MaterialPolypropylene SMS Fabric
ColorWhite
Closure TypeTies
DisposableSingle-use
Latex FreeYes
Model36072
SterilitySterile
Certifications/ComplianceType 6, Category III PPE
Quantity75/Cs.
IncludesPull down ties, stretch-fit elastic, blue donning line
For Use With (Application)Laboratory Protection
Hood TypeIntegrated
Cleanroom ClassClass 100 (ISO 5)
Unit SizeCase