Aerosolv™ 360 System
Aerosolv™ 360 System
Actual product may vary
Aerosolv™ 360 System
Justrite MFG LLC Aerosolv™ 360 System is a premium puncturing unit for recycling aerosol cans
Have Questions?
Catalog number KLC28100
Price (SGD)
-
  • Enclosed system—dual seal chamber eliminates the potential for content spray back, prevents VOC release and reduces gunk build up
  • High-efficiency combination coalescing carbon filter captures odors, and prevents potentially harmful VOCs from being released into the air—extended filter life reduces change outs to save time and money
  • Alignment cone and two-way pressure controlled, non-sparking vent pin offers the most efficient point of puncture—greatly reduces cleanup and minimizes maintenance
  • Intuitive, ergonomic activation wheel design is easy to use, and with no lever handle extending over the side of the drum, removes the chance for snagged clothing or tipped drums
  • Ideal for high volume applications - safely converts spent aerosol cans from solid hazardous waste to a non-hazardous, recyclable material.
  • Threading directly into a standard 2-in (51-mm) bung on a 30- (110-liter) or 55-gallon (200-liter) drum, the housing accommodates a wide variety of the most common sized cans, including domed mini, standard, and jumbo.
  • Sleek design with no protruding lever handle—mechanical activation wheel with rack and pinion gear increases torque making can processing easy and fast.
  • Integrated lock out system prevents unauthorized use and tamper-proof retention plate secures the unit to the drum.
  • Non-sparking puncture pin pierces the can and directs the liquid flow straight into the drum for a quick, controlled evacuation.
  • Non-stick coating on housing minimizes static transfer to the unit while at the same time reduces clean up.
  • Attach the two-stage high-efficiency combination filter to the ¾-in (19-mm) bung to capture VOCs from any remaining propellants.
  • Replaceable, proprietary polymer coalescer in filter base captures the residual liquid in the gas before it reaches the activated carbon, helping extend the life of the filter.
  • The top activated carbon filter cartridge features adsorption indicator which reacts to the adsorption of VOCs to indicate when the filter cartridge is ready for replacement.
  • Built-in counter tracks system use for maintenance.
  • Includes safety goggles and an anti-static ground wire with alligator clip.
  • One 55-gallon (200-liter) drum holds approximately 4,000 spent aerosol cans.
  • Punctured, discharged, cans do not meet the definition of hazardous waste per EPA 40 CFR 261.23(a)(6), and thus can be treated as recyclable scrap metal.
Specifications
Certifications/ComplianceEPA compliant
ColorBlack, Green
Quantity1/Pk.
AutoclavableNot Autoclavable
IncludesPremium puncturing unit, high-efficiency filter, wire, and goggles.
For Use With (Application)Recycling aerosol cans
For Use With (Equipment)55 gallon drum
Height (English)17 in.
Unit SizeCase