Coated GC Packing, Phase 0.2 Carbowax 1500, Matrix 80/100 Carbopack C Support
Coated GC Packing, Phase 0.2 Carbowax 1500, Matrix 80/100 Carbopack C Support
Actual product may vary
Coated GC Packing, Phase 0.2 Carbowax 1500, Matrix 80/100 Carbopack C Support
Ready-to-use coated packing for those wanting to pack their own columns.
Have Questions?
Catalog number KLC11827
Price (SGD)
668.00
CS
Add to cart
Price (SGD)
668.00
CS
Add to cart
The choice of coating material is based on the stationary phase of the GC.
Specifications
PackagingBottle
Quantity15 g
For Use With (Application)Gas Chromatography (GC)
Unit SizeCS