Coated GC Packing, Phase 0.2 Carbowax 1500, Matrix 60/80 Carbopack C Support
Coated GC Packing, Phase 0.2 Carbowax 1500, Matrix 60/80 Carbopack C Support
Actual product may vary
Coated GC Packing, Phase 0.2 Carbowax 1500, Matrix 60/80 Carbopack C Support
Ready-to-use coated packing for those wanting to pack their own columns.
Have Questions?
Catalog number KLC11826
Price (SGD)
-
The choice of coating material is based on the stationary phase of the GC.
Specifications
PackagingBottle
Quantity15 g
For Use With (Application)Gas Chromatography (GC)
Unit SizeCS