Kimtech™ A8 Lab Coats, Traditional Collar
Actual product may vary
Kimtech™ A8 Lab Coats, Traditional Collar
Kimtech™ A8 Lab Coats, Traditional Collar
Kimtech™ A8 Lab Coats, Traditional Collar
Non-woven SMS lab coat provides comfort and protection from light fluid contact.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSize
KLC10120Small
KLC10122Large
KLC10121Medium
FIS19-0430612X-Large
KLC10123X-Large
Catalog number KLC10120
Price (SGD)
164.00
Case
Size:
Small
Price (SGD)
164.00
Case
  • Protective three-layer white SMS fabric resistant to blood strike-through for various levels of exposure
  • Feature traditional collar, knit cuffs, four snaps, two reinforced pockets, and size/name label
OSHA requires the employer to evaluate the task and type of exposure anticipated and select the appropriate clothing that prevents contamination of an employee's skin or clothing by blood or other potentially infectious materials
Specifications
Product TypeGeneral Purpose Lab Coat
SizeSmall
GenderUnisex
MaterialPlastic, Plastic
ColorRed on White, Red on White
Closure TypeSnaps
Cuff StyleKnit
Quantity1 Ea.
SterilityNon-sterile
No. of Pockets2
Collar TypeTraditional
Diameter (English)10 in.
Model10120
Mounting TypeCorner Holes
For Use With (Application)Communication
Size (English) Chest30 to 34 in.
Height (English)7 in.
Thickness3-ply
Width (English)10 in.
LanguagesEnglish
Dimensions (L x W)7 x 10 in.
Displayed TextDANGER Gas Cylinder Storage Area
Construction3-ply SMS
Unit SizeCase