Kimble™ KIMAX™ Griffin Beakers
Kimble™ KIMAX™ Griffin Beakers
Actual product may vary
Kimble™ KIMAX™ Griffin Beakers
With thick, beaded, slightly flared top
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
KIM1400015001500 mL
FIS02-539C10 mL
FIS02-539-120 mL
FIS02-539F30 mL
FIS02-539G50 mL
FIS02-539M600 mL
FIS02-539P1000 mL
FIS02-539Q1500 mL
FIS02-539R2000 mL
FIS02-539T4000 mL
Catalog number KIM140001500
Price (SGD)
-
Capacity (Metric):
1500 mL
With thick, beaded, slightly flared top
  • With pouring spout
  • Matte-finish enameled marking spot
  • 20 to 4000mL sizes have white scales for approximate volume
  • Accurate to ±5%
  • Uniform, sturdy walls
Specifications
Accuracy±5%
Capacity (English)50.72 oz.
Capacity (Metric)1500 mL
MaterialBorosilicate Glass
For Use With (Application)For Measuring and Mixing Liquids
Graduation Range200 to 1400 mL
IncludesSpout
Increments200 mL
Label TypeMatte Finish Enamel Marking
Quantity4 Pk.
TypeBeaker
DisposableReusable
GraduatedYes
Certifications/ComplianceASTM E960, Type I
ScaleDual White Scale
StyleGriffin
Unit SizePK