Texwipe™ Low TOC Alpha™ Swab
Texwipe™ Low TOC Alpha™ Swab
Actual product may vary
Texwipe™ Low TOC Alpha™ Swab
Constructed to exacting and consistent tolerances without the use of adhesives for superior performance. Texwipe™ Low TOC Alpha™ Swabs are cleeaned using a proprietary process to ensure suitability for use in Total Organic Carbon (TOC) analysis as part of a cleaning validation protocol.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)
ITWTX761K162.3 mm
FIS19-034-963162.3 mm
FIS19-034-962127.5 mm
ITWTX714K127.5 mm
Catalog number ITWTX761K
Price (SGD)
-
Length (Metric):
162.3 mm
Constructed to exacting and consistent tolerances without the use of adhesives for superior performance. Texwipe™ Low TOC Alpha™ Swabs are cleeaned using a proprietary process to ensure suitability for use in Total Organic Carbon (TOC) analysis as part of a cleaning validation protocol.
  • Certified; Cleaned to a level of <50μg/L TOC ensures consistency in swab background contribution
  • Ideal for use in cleaning validation of equipment used in manufacture of APIs, excipients and drugs
  • Double-layered, double-knit polyester head provides excellent recovery rates by entrapping samples in knit structure without adhering to sample, allowing complete release into diluent
  • Large head on rigid, break-away handle provides firm support for sampling broad areas and flat surfaces and allows head to be placed in vial with minimal handling and contamination
  • 100% polypropylene handle ensures no additional contaminants are introduced and provides excellent chemical resistance
  • No background interference from other contaminants
  • Autoclavable for use in sterile environments and processes
Specifications
DescriptionLong Handle
ConstructionFlexible head paddle, long handle
For Use With (Application)Surface sampling for use in cleaning validation protocols, precision cleaning of hard-to-reach areas or small spaces,
Handle ColorLight Green
Diameter (Metric) Handle3.2 mm
Length (Metric)162.3 mm
Length (Metric) Handle145.5 mm
Thickness (Metric)2.8 mm
Length (Metric) Tip16.8 mm
Width (Metric) Tip6.8 mm
Adhesive TypeThermal
Handle MaterialPolypropylene
Tip MaterialKnit Polyester
Unit SizeCase