Check Order Status
TexMop™ Aluminum Handles for Cleanroom Mops
Actual product may vary
TexMop™ Aluminum Handles for Cleanroom Mops
For use with AlphaMop™, Mini AlphaMop™/Isolator Cleaning Tool and ClipperMop™.
Have Questions?
Catalog Number
Length (English)
ITWTX7183
55 to 92 in.
Catalog number
ITWTX7183
Price (SGD)
-
Length (English):
55 to 92 in.
Telescoping type
Blue color
Use with TexConnect™ adapter to attach to screw-based mop heads
Specifications
Product Type
Cleanroom Mop Handle
Color
Blue
Length (English)
55 to 92 in.
Length (Metric)
140 to 233.6 cm
Quantity
1/Pk.
For Use With (Equipment)
AlphaMop, Mini AlphaMop, Isolator Cleaning Tool, ClipperMop
Material
Aluminum