TexMop TX7115 Stainless Steel Mop Head
TexMop TX7115 Stainless Steel Mop Head
Actual product may vary
TexMop TX7115 Stainless Steel Mop Head
TexMop TX7115 Stainless Steel Mop Head is a stainless steel flat mop for cleaning and disinfecting surfaces such as ceilings, floors, walls, isolators, confined and hard-to-reach areas. Use with any 9 x 9 in. wiper with a mop pad.
Have Questions?
Catalog NumberDimensions
ITWTX7115LxWxH: 8 x 4 x 2 in. (20 x 10 x 5 cm)
Catalog number ITWTX7115
Price (SGD)
-
Dimensions:
LxWxH: 8 x 4 x 2 in. (20 x 10 x 5 cm)
  • TexMop TX7115 is a stainless steel, low profile, swivel head, flat mop head ideal for cleaning surfaces such as ceilings, floors, walls, confined and hard-to-reach areas.
  • TexMop can be used with any 9 x 9 in. (23 x 23 cm) wipers with a mop pad.
  • Wipers easily fit over the head into the TexStar connector permitting quick changes.
  • TX7115 is suitable for applying and removing solutions and disinfectants.
  • TexMop can be also used with Revolve 2-in-1 Integrated Sustainable Covers. These 100% polyester fabric covers are integrated with a polyester pad. These 2-in-1 covers do not require an additional mop pad to be used.
  • The TexStar Connector system adapter allows for easy and frequent wiper replacement for adherence to strict cleaning protocols
  • Stainless steel with an ergonomic, stable design
  • Replacement pads and fasteners available
  • Autoclave safe
Specifications
DimensionsLxWxH: 8 x 4 x 2 in. (20 x 10 x 5 cm)
Product TypeCleaning Tool
Head TypeFlat, Low Profile, Swivel Head
ColorGray
Length (English)4 in.
SterilityNon-sterile
Length (Metric)10 cm
AutoclavableAutoclavable
Height (Metric)5 cm
Height (English)2 in.
Width (Metric)20 cm
Width (English)8 in.
DisposableReusable
Quantity1 Ea.
Includes1 Mop Head, 10 Polyester Pads
For Use With (Equipment)Floors, Walls, Ceilings, Isolators, Hard-to-reach Areas
For Use With (Application)Cleaning and Applying Disinfectants
Color-codedNo
MaterialStainless Steel, Polypropylene
Unit SizeCase