ClipperMop™ Mop
ClipperMop™ Mop
Actual product may vary
ClipperMop™ Mop
For use with wet, dry, or sterile wipers
Have Questions?
Catalog NumberDimensions
ITWTX710311 x 4 in. Head Assembly
Catalog number ITWTX7103
Price (SGD)
-
Dimensions:
11 x 4 in. Head Assembly
For use with wet, dry, or sterile wipers
  • Compatible with protocols for ISO Class 3 to 8 cleanrooms, sterile and aseptic areas
  • Designed to use standard 9 × 9 in. (23 × 23cm) and 12 × 12 in. (30.5 × 30.5cm) cleanroom wipers—eliminate need to qualify and inventory additional wipers or mop covers
  • Patent-pending mophead utilizes nonabrasive polyethylene clips to securely fasten one or multiple wipers to the head
  • Wiper attachment method does not release particles, fibers, or dirt
  • System can be sterilized by gamma irradiation, chemicals, vaporized hydrogen peroxide (VHP), or ethylene oxide (EtO)
Specifications
Dimensions11 x 4 in. Head Assembly
Product TypeCleanroom Mop
SterilitySterile
IncludesHead Assembly, 30 to 53 in. Telescoping Handle
Certifications/ComplianceISO Class 3 to 8
Unit SizeCase