CleanTip™ Swabs: Foam Series
CleanTip™ Swabs: Foam Series
Actual product may vary
CleanTip™ Swabs: Foam Series
CleanTip™ Swabs: Foam Series
CleanTip™ Swabs: Foam Series
Certified to be silicone-free
Have Questions?
Catalog NumberTip StyleLength (Metric) HandleDiameter (Metric) Tip
ITWTX708AFlexible105.8 mm19.2 mm
Catalog number ITWTX708A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Tip Style:
Flexible
Length (Metric) Handle:
105.8 mm
Diameter (Metric) Tip:
19.2 mm
Certified to be silicone-free
High performance for applications requiring sorbency, low particle generation and solvent resistance.
  • High-grade, closed-cell polyurethane foam offers high performance for applications requiring sorbency, low particle generation, and solvent resistance
  • Softness and low abrasion protect delicate components
  • Foam head on flexible and rigid paddle, both with chemical-resistant polypropylene handles
  • Constructed from the cleanest materials available and manufactured in a cleanroom using state-of-the-art processes and equipment
  • High-precision automated process guarantees consistent performance, sizing and cleanliness
  • Silicone-free Bag-Within-A-Bag™ packaging consists of a solvent-resistant, thermally sealed exterior bag that protects the inner bags containing the swabs
  • Each outer bag is individually lot-coded for easy traceability and quality control
Specifications
ConstructionRigid head paddle, long, easy-grip handle
Diameter (English) Handle0.26 in.
Length (English)5.205 in.
For Use With (Application)Applying and removing lubricants, adhesives and other solutions in a critical clean environment, scrubbing recessed areas, removal of excess materials, debris, cleaning intersecting surfaces and joints
Handle ColorLight Green
Product TypeSwab
Diameter (Metric) Handle6.6 mm
Length (Metric)132.2 mm
Length (English) Handle4.154 in.
Length (Metric) Handle105.8 mm
MaterialFoam
ShapeCircular
Thickness (Metric)9.4 mm
Length (English) Tip1.04 in.
Handle TypeLong, easy-grip
Length (Metric) Tip26.4 mm
Width (Metric) Tip6.6 mm
Width (English) Tip0.12 in.
Diameter (Metric) Tip19.2 mm
Diameter (English) Tip0.756 in.
Adhesive TypeThermal
Handle MaterialPolypropylene
Tip StyleFlexible
Tip MaterialPolyurethane Foam
Unit SizeCase