TechniCloth™ Nonwoven Dry Wipers, ISO Class 6/7
TechniCloth™ Nonwoven Dry Wipers, ISO Class 6/7
Actual product may vary
TechniCloth™ Nonwoven Dry Wipers, ISO Class 6/7
For ISO Class 6 and 7 applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)
ITWTX61212 x 12 in.
FIS18-315D12 x 12 in.
FIS18-315A4 x 4 in. (10.16 x 10.16 cm)
ITWTX6099 x 9 in. (22.86 x 22.86 cm)
Catalog number ITWTX612
Price (SGD)
-
Dimensions (L x W):
12 x 12 in.
For ISO Class 6 and 7 applications
Economical all-purpose nonwoven wipers made from a blend of 55% cellulose and 45% polyester—combine the absorbency of natural fiber with the cleanliness and strength of synthetic fiber.
  • Hydroentanglement process uses jets of water to blend fibers, eliminating need for chemical binders
  • Provide excellent bidirectional strength and very low solvent extractables levels
  • Offer superior wet and dry strength, solvent-compatibility and acid resistance
  • Will not scratch or abrade delicate precision components
Specifications
Product TypeDry Wipe
Length (English)12 in.
Length (Metric)30.5 cm
Width (English)12 in.
Width (Metric)30.5 cm
MaterialPolyester Cellulose
AutoclavableAutoclavable
Basis Weight69
Certifications/ComplianceISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Dimensions (L x W)12 x 12 in.
For Use With (Application)Designed for general purpose wiping where economy and cleanliness are of critical importance, ideal for applications where high absorbency and low extractables are prime considerations
Unit SizeCase