TexBond™ 20 Cleanroom Bond Paper
TexBond™ 20 Cleanroom Bond Paper
Actual product may vary
TexBond™ 20 Cleanroom Bond Paper
Cleanroom bond paper with exceptional contamination control characteristics needed in critical environments. It has been especially formulated to address the needs of users requiring a paper that retains low ion and particle levels with significantly improved copying and writing qualities.
Have Questions?
Catalog NumberColor
ITWTX5711Blue
Catalog number ITWTX5711
Price (SGD)
-
Color:
Blue
  • Possesses both strength and excellent heat resistance, making it ideal for use in laser printers and photocopiers
  • Very low particle generation
  • Very low ion levels
  • High opacity rating
  • Precision cut edges
  • Cleanroom packaged Benefits
  • Exceptional copying ability
  • Laser printer and photocopier compatible
  • Clear reproductions
  • Autoclavable for use in sterile environments
  • Photocopying, ink jet and laser printing
  • Offset printing
  • Cleanroom manuals, work instructions, note taking and data transfer
  • Color coded carriers
Specifications
ColorBlue
Dimensions (L x W)210 x 297 mm
Quantity250 Sheets/Pk., 10 Packs/Cs.
Unit SizeCase