Sterile 70% Isopropanol
Sterile 70% Isopropanol
Actual product may vary
Sterile 70% Isopropanol
Evaporates with extremely low residue
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (English)Container Type
ITWTX32901 gal.
FIS19-130-386016 oz.Trigger Spray Bottle
ITWTX327016 oz.Trigger Spray Bottle
ITWTX327332 oz.Trigger spray bottle
Catalog number ITWTX3290
Price (SGD)
-
Volume (English):
1 gal.
Sterile solution contains 70% by volume USP grade isopropyl alcohol with 30% USP purified water. Solution is 0.2μm filtered, filled into cleaned containers, double bagged and packaged in sealed polybag case liner, and gamma-irradiated to a sterility level of 10-6.
  • Evaporates with extremely low residues
  • Eliminates mixing, filtration, sterilization, processing and QC expense of in-house production of sterile alcohol/water blends
  • Each container is lot coded with an expiration date
Specifications
FormLiquid
SterilitySterile
AppearanceClear
Certifications/ComplianceUSP
Composition70% USP Isopropyl Alcohol and 30% USP Purified Water
Concentration70%
Filtered0.2 μm
For Use With (Application)Surface cleaning, wide down for pass through to controlled environments, eliminates mixing, filtration, sterilization, processing and QC expense of in-house production of sterile Alcohol/Water blends
IncludesCertificates of processing, analysis and compliance
Volume (English)1 gal.
Unit SizeCase