Sterile TechniSat™ Presaturated Wipers
Sterile TechniSat™ Presaturated Wipers
Actual product may vary
Sterile TechniSat™ Presaturated Wipers
For ISO Class 6 applications
Have Questions?
Catalog NumberPackaging Quantity
ITWTX32141000 Wipers total/Cs.
Catalog number ITWTX3214
Price (SGD)
-
Packaging Quantity:
1000 Wipers total/Cs.
For ISO Class 6 applications
  • Binder-free hydroentangled polyester/cellulose wiper presaturated with USP-grade 70% isopropyl alcohol 30% deionized water, filtered through 0.2μm filter
  • Gamma irradiated to 10-6 Sterility Assurance Level in accordance with Association for Advancement of Medical Instrumentation
  • Low levels of solvent extractables and particle generation
Specifications
Product TypePre-saturated Wiper
Length (English)11 in.
Length (Metric)28 cm
Width (English)11 in.
Width (Metric)23 cm
MaterialPolyester Cellulose
Dimensions (L x W)11 x 9 in. (28 x 23 cm)
For Use With (Application)Designed for cleaning and polishing equipment and environmental surfaces during and following the production flow
Packaging Quantity1000 Wipers total/Cs.
Saturation Solution70% Isopropyl Alcohol, 30% Deionised Water