SterileWipe™ Sterile Dry Wipers
SterileWipe™ Sterile Dry Wipers
Actual product may vary
SterileWipe™ Sterile Dry Wipers
For aseptic cleanrooms
Have Questions?
Catalog NumberDimensions (L x W)Material
ITWTX32119 x 9 in. (22.86 x 22.86 cm)Polyester
Catalog number ITWTX3211
Price (SGD)
-
Dimensions (L x W):
9 x 9 in. (22.86 x 22.86 cm)
Material:
Polyester
Specifically developed for maximum control of particulate contamination in sterile environments.
SterileWipe LP
  • Knitted from 100% continuous filament polyester
  • Double-knit, no-run interlock construction ensures maximum abrasion resistance and low particle values
  • Unique knit pattern also provides greater surface area for higher sorbency and increased wiping efficiency
  • Soft and nonabrasive; ideal for wiping sensitive surfaces
  • Basis weight: 155g/m2
SterileWipe LP 10
  • Sealed-border wiper with no-run interlock double-knit construction of 100% continuous filament polyester
  • All four edges thermally sealed to prevent release of fibers and particulates
  • Designed for use in an ISO Class 4 cleanroom to further ensure ultralow particulate and extractable levels
  • Ideal for wiping and spill control Basis weight: 135g/m2
SterileWipe HS II
  • Blend of 55% cellulose and 45% polyester combines the absorbency of natural fiber with the strength of synthetic fiber
  • Ideal for applications where bacteriological contamination, particulates and extractables are concerns
  • Appropriate for use in aseptic environments of the pharmaceutical, medical device and biomedical industries
  • Basis weight: 67g/m2
Specifications
Product TypeDry Cleanroom Wiper
Length (English)9 in.
Length (Metric)22.9 cm
Width (English)9 in.
Width (Metric)22.9 cm
MaterialPolyester
Basis Weight155
Certifications/ComplianceISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
DescriptionAlphaWipe™ Dry Cleanroom Wipers
Dimensions (L x W)9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application)For aseptic cleanrooms