Vectra™ QuanSat™ Cleanroom Wipers
Vectra™ QuanSat™ Cleanroom Wipers
Actual product may vary
Vectra™ QuanSat™ Cleanroom Wipers
For use in ISO Class 3 to 6 applications
Have Questions?
Catalog NumberSaturation Solution
ITWTX10866% Isopropyl Alcohol
Catalog number ITWTX1086
Price (SGD)
-
Saturation Solution:
6% Isopropyl Alcohol
  • Vectra QuanSat™ thermally sealed-border wiper presaturated with various concentrations of semi-conductor-grade isopropyl alcohol
  • Economical 100% synthetic continuous filament, double-knit polyester
  • Superior strength for excellent wiping efficiency and abrasion resistance
  • Ultralow particle levels and low extractable levels
Specifications
Product TypeCleanroom Wipe
Length (English)9 in.
Length (Metric)22.9 cm
Width (English)9 in.
Width (Metric)22.9 cm
MaterialPolyester
Basis Weight109
Certifications/ComplianceISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
EdgesSealed
Dimensions (L x W)9 x 9 in. (22.86 x 22.86 cm)
For Use With (Application)Cleanroom
Saturation Solution6% Isopropyl Alcohol
Unit SizeCase