Polysat™ Presaturated Polypropylene Wipers
Polysat™ Presaturated Polypropylene Wipers
Actual product may vary
Polysat™ Presaturated Polypropylene Wipers
For ISO Class 6 to 7 applications
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)SterilityPackaging Quantity
ITWTX105111 x 9 in. (28 x 23 cm)Non-sterile1200/Cs.
ITWTX104011 x 7 in. (27.9 x 17.7 cm)Non-sterile800/Cs.
Catalog number ITWTX1051
Price (SGD)
-
Dimensions (L x W):
11 x 9 in. (28 x 23 cm)
Sterility:
Non-sterile
Packaging Quantity:
1200/Cs.
  • Constructed from a soft, 100% meltblown polypropylene substrate
  • Presaturated with 70% isopropyl alcohol/30% deionized water solution
  • Low particle generation and low levels of solvent extractables
Specifications
Product TypePre-wetted Wipe
Length (English)11 in.
Length (Metric)27.9 cm
Width (English)11 in.
Width (Metric)22.8 cm
MaterialPolypropylene
Basis Weight35
Certifications/ComplianceISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
Dimensions (L x W)11 x 9 in. (28 x 23 cm)
For Use With (Application)Designed for cleaning equipment and other environmental surfaces during and following the production flow, suitable for wiping the exterior of production equipment in controlled environments
Packaging Quantity1200/Cs.
Saturation Solution70% Isopropyl Alcohol, 30% Deionised Water
SterilityNon-sterile
Includes24 Packs of 50 wipers
Unit SizeCase