CleanFoam Sterile Foam Swabs
CleanFoam Sterile Foam Swabs
Actual product may vary
CleanFoam Sterile Foam Swabs
Texwipe CleanFoam Sterile Foam Swabs, Length: 5 in., 132.2 mm, Width Head: 0.78 in., 19.7 mm, Handle Type: Polypropylene, Polyurethane, Recommended Applications: Safety
Have Questions?
Catalog NumberLength (English)Width (English) Head
ITWSTX708A5 in.0.78 in.
Catalog number ITWSTX708A
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Length (English):
5 in.
Width (English) Head:
0.78 in.
  • Texwipe CleanFoam Sterile Foam Swabs
  • Length: 5 in., 132.2 mm
  • Width Head: 0.78 in., 19.7 mm
  • Handle Type: Polypropylene, Polyurethane
  • Recommended Applications: Safety
  • Environmental monitoring
  • Cleaning with compatible solutions and solvents
  • Picking up fine powders
  • Surface sampling
  • Collection and transport of bacteriological specimens
  • Specifications
    Length (English)5 in.
    Product TypeSwab
    Length (Metric)132.2 mm
    MaterialPolyurethane
    Width (English) Head0.78 in.
    Width (Metric) Head19.7 mm
    Handle TypePolypropylene
    Adhesive TypeComplete Thermal Bond
    Handle MaterialPolypropylene
    Tip StyleCircular
    Tip MaterialPolyurethane
    Unit SizeCase