Spark-Free Interior, Lab Freezer
Spark-Free Interior, Lab Freezer
Actual product may vary
Spark-Free Interior, Lab Freezer
For secure, accurate and long-lasting freezer storage of temperature-sensitive products, the Spark-Free Interior, Lab Freezer is fully-equipped for lab applications with Liebherr build-quality you can rely on. EN 60068 temperature mapping and SmartMonitoring options.
Have Questions?
Catalog NumberCapacity (Metric)Temperature RangeStyleModel
ITSLGUEX1500139 L-9°C to -26°CTabletopLGUex1500
Catalog number ITSLGUEX1500
Price (SGD)
5,133.00
EA
Add to cart
Capacity (Metric):
139 L
Temperature Range:
-9°C to -26°C
Style:
Tabletop
Model:
LGUex1500
Price (SGD)
5,133.00
EA
Add to cart
  • ATEX-certified spark-free interior
  • Can be validated to FD X15-140, GLP-compliant.
  • Manual defrost.
  • White-coated steel exterior finish.
  • Moulded inner liner with coved corners.
  • Door with lock, reversible on site.
  • Self-closing door.
  • Access port Ø 15mm for external temperature probes to each compartment.
  • Access port Ø 30mm for validation on site, sited on the side wall at 1/3 of the height, to each compartment
  • Electronic controller with digital display with 0,1°C resolution.
  • Digital keypad lock with PIN code.
  • Hi-low optical and acoustic temperature alarms with adjustable thresholds.
  • Volt-free contact for connection to a remote alarm system.
  • Alarm log with memory of the last 3 temperature alarms with time, date and duration.
  • Alarm log with memory of the last 3 power failure alarms with time, date and duration.
  • Min - max temperature memory.
  • RS 485 interface for external temperature monitoring and centralised alarm systems.
Specifications
TypeFreezer
Capacity (Metric)139 L
Temperature Range-9°C to -26°C
Temperature Resolution0.1°C
Temperature StabilityGradient 6.6°C
Door StyleSolid
No. of Doors1
Door MaterialSteel
InsulationHDPE
Exterior FinishWhite Coated Steel
Capacity (Metric) Shelf40 kg
No. of Drawers3
Certifications/ComplianceATEX, CE, ISO 14001, ISO 9001
StyleTabletop
CastersNo
ColorWhite
Control TypeDigital
DefrostManual
ModelLGUex1500
Depth (Metric) Exterior61.5 cm
Depth (Metric) Interior45 cm
Height (Metric) Exterior82 cm
Height (Metric) Interior66.3 cm
Width (Metric) Exterior59.7 cm
Width (Metric) Interior45.4 cm
Monitoring OptionsSmartCoolingHub
Data OutputsRS 485
Amperage1.0 A
Voltage220/240 V
Frequency50 Hz
Wattage100 w
Energy Usage338 kWh/a
RefrigerantR 600a
Shipping Weight (Metric)45 kg
Weight (Metric)42 kg
Unit SizeEA