Tall Lab Fridge/Freezer
Tall Lab Fridge/Freezer
Actual product may vary
Tall Lab Fridge/Freezer
Tall Lab Fridge/Freezer
Tall Lab Fridge/Freezer
Tall Lab Fridge/Freezer
Tall Lab Fridge/Freezer
For secure, accurate and long-lasting fridge-freezer storage of temperature-sensitive products, the Liebherr™ Tall Lab Fridge/Freezer is fully-equipped for lab applications with Liebherr build-quality you can rely on. EN 60068 temperature mapping and SmartMonitoring options.
Have Questions?
Catalog NumberModel
ITSLCV4010LCV4010
Catalog number ITSLCV4010
Price (SGD)
-
Model:
LCV4010
  • Upright lab fridge-freezer, reach-in, 1-section, width 600mm, gross capacity 254/107 L.
  • Temperature ranges +3 / +16°C and -9/-30°C, 3 adjustable shelves, 3 freezer drawers, white exterior, commercial-grade moulded polystyrol interior, fridge section: fan-assisted refrigeration, freezer section: static refrigeration, manual defrost.
  • Climate class 7, refrigerant R 600a, connection rating 1,5A, 230v/50/1-ph, energy consumption 657 kWh/a
  • 240/105 litre net capacity
  • 2 separate refrigeration systems
  • Freezer drawers: easy product handling, maximum energy efficiency and minimum temperature fluctuation
  • Digital controller with 0,1°C resolution, password keypad lock, RS 485 interface and volt-free alarm contact
  • Access port for external temperature probe, additional safety thermostat
  • Alarm log of last 3 events with min/max temperatures, duration etc.
  • Door locks, reversible hinges, self-closing doors
Specifications
Capacity (Metric) Freezer107 L
Capacity (Metric) Refrigerator254 L
Temperature Range (Freezer)-9°C to -30°C
Temperature Range (Refrigerator)3°C to 16°C
Temperature Resolution0.1°C
Temperature StabilityGradient 5.2°C / 7.9°C
Door StyleSolid
No. of Doors2
DrawersTransparent (Freezer)
No. of Drawers3
Shelves4
Capacity (Metric) Shelf45 kg
DefrostAutomatic (Refrigerator), Manual (Freezer)
Monitoring OptionsSmartCoolingHub
Data OutputsRS 485
RefrigerantR 600a
Exterior FinishWhite Epoxy-coated Steel
Insulation55 mm HDPE (refrigerator)/70 mm HDPE (freezer)
InteriorABS Plastic
Certifications/ComplianceCE, COFRAC, ISO 14001, ISO 9001, QCPP (Pharmacy guild of Australia): LKV3912, LKV3910, LKUV1612, LKUV1610
StyleTall
CastersNo
ColorWhite
Condenser TypeVentilated (fridge), static (freezer)
ModelLCV4010
Warranty24 Months
Dimensions (L x W x H) Exterior60 cmL x 61.5 cmD x 200 cmH
Depth (Metric) Exterior61.5 cm
Depth (Metric) Interior44.1 cm
Height (Metric) Exterior200.3 cm
Height (Metric) Freezer59.7 cm
Height (Metric) Interior110.5 cm
Height (Metric) Refrigerator110.5 cm
Width (Metric) Exterior59.7 cm
Width (Metric) Freezer43.1 cm
Width (Metric) Interior44.0 cm
Width (Metric) Refrigerator45.5 cm
Energy Usage657 kWh/a
Amperage1.5 A
Voltage220/240 V
Frequency50 Hz
Wattage260 w
Shipping Weight (Metric)90 kg
Weight (Metric)84 kg
Unit SizeEA