Hotplate Stirrer, 320°C, Stainless Steel
Hotplate Stirrer, 320°C, Stainless Steel
Actual product may vary
Hotplate Stirrer, 320°C, Stainless Steel
Shape: Round, Size: 155 mm dia., Plug Type: EU-Style
Have Questions?
Catalog NumberPlug Type
IKA5019802EU
Catalog number IKA5019802
Price (SGD)
2,917.00
EA
Add to cart
Plug Type:
EU
Price (SGD)
2,917.00
EA
Add to cart
Shape: Round; Size: 155 mm dia.; Plug Type: EU-Style
Specifications
Top Plate MaterialStainless Steel
Voltage220/240 V
IncludesM10 Thread
Size155 mm dia.
Height (Metric)246 mm
Temperature (Metric)Ambient to 320°C
Width (Metric)90 mm
Length (Metric)160 mm
Wattage600 W
Weight (English)4.4 lb.
Weight (Metric)2 kg
TypeStirring Hotplate
ShapeRound
Plug TypeEU
No. of Positions1
Max. Container Size15 L
Unit SizeEA