Magnetic Stirrer
Magnetic Stirrer
Actual product may vary
Magnetic Stirrer
Designed for synchronous stirring without heat. IKA Magnetic Stirrer includes magnetic coil technology to provide noiseless and consistent stirring on all positions. Speed remains constant, even when load changes.
Have Questions?
Catalog NumberModelStirring PositionsTop Plate Surface Area (Metric)
IKA3692500RO 1515280 x 470 mm
Catalog number IKA3692500
Price (SGD)
9,384.00
EA
Add to cart
Model:
RO 15
Stirring Positions:
15
Top Plate Surface Area (Metric):
280 x 470 mm
Price (SGD)
9,384.00
EA
Add to cart
Digital magnetic stirrer without heating, designed for synchronous stirring. Magnetic coil technology provides noiseless and consistent stirring on all positions. Speed remains constant, even when load changes. Closed and compact design allows easy cleaning and protects the equipment against the penetration of liquids. Additionally, a removable transparent anti-slip mat is included.
  • Adjustable reverse operation
  • Wear free magnetic coils
  • Error code display
  • Easy operation with touch keypad
  • Suitable for continuous operation
  • Speed range: 0 to 1200 rpm
Specifications
Speed0 to 1200 rpm
Stirring Positions15
Top Plate MaterialStainless Steel
Voltage100/240 V
Wattage40 w
Height (Metric)60 mm
Top Plate Surface Area (Metric)280 x 470 mm
Width (English)11 in.
Width (Metric)280 mm
Amperage1500 mA
Frequency50/60 Hz
ModelRO 15
Temperature Range (Metric)5°C to 40°C
TypeRO 15 Magnetic Stirrer
Weight (Metric)7 kg
Unit SizeEA