R 182 Boss Head Clamp
R 182 Boss Head Clamp
Actual product may vary
R 182 Boss Head Clamp
Secure joints with this clamp. IKA™ R 182 Boss Head Clamp features a sturdy, cast aluminum construction and cross sleeve design. Ideal for a variety of applications.
Have Questions?
Catalog NumberDiameter (Metric)
IKA26577006 to 16 mm
Catalog number IKA2657700
Price (SGD)
196.00
EA
Add to cart
Diameter (Metric):
6 to 16 mm
Price (SGD)
196.00
EA
Add to cart
  • Clamping range, stand: 6 to 16mm
  • Clamping range, extension arm: 6 to 16mm
Specifications
Product TypeBoss Head Clamp
Diameter (Metric)6 to 16 mm
MaterialCast Aluminum
Packaging Quantity1 Ea.
Unit SizeEA