GammaGuard CE™ Sterile Cleanroom Sleeves
GammaGuard CE™ Sterile Cleanroom Sleeves
Actual product may vary
GammaGuard CE™ Sterile Cleanroom Sleeves
GammaGuard CE™ Sterile Cleanroom Sleeves
GammaGuard CE™ Sterile Cleanroom Sleeves
GammaGuard CE™ Sterile Cleanroom Sleeves are made of a comfortable fabric that protects against various liquids and particulates.
Have Questions?
Catalog number IENCE11065CIS
Price (SGD)
446.00
CS
Add to cart
Price (SGD)
446.00
CS
Add to cart
Sterile sleeves are made of a comfortable fabric that protects against various liquids and particulates. Each sleeve is individually packaged and sterilized to a 10-6 sterility assurance level (SAL) preventing workplace contamination and cross-contamination. The low-linting fabric helps reduce the release of unwanted particulates to prevent product damage and workplace contamination. Sleeves are designed with tunnelized elastic, providing a more secure closure and stronger seams. This also contains raw edges that can create and release additional particulates. Sleeves are tear-resistant and made in an ISO-certified facility to ensure the highest quality.
  • Sterile-Don™ packaging protects against contamination during donning
  • GammaGuard CE™ helps keep the wearer cooler vs many competitive fabrics
  • Generous 21.5 in. length provides excellent arm coverage
  • Meets high manufacturing standards - made in an ISO-certified facility
  • Certificate of sterility included in each case
  • The tear-resistant fabric ensures your safety in challenging environments
Specifications
SizeStandard
MaterialMicroporous Fabric
ColorWhite
SterilitySterile
Seam TypeSerged
Cuff StyleElastic
Latex FreeYes
ModelCE11065CIS
Length (English)21.5 in.
Length (Metric)54.6 cm
PackagingIndividually Packaged
Unit SizeCS