Lindberg/Blue M™ 1200°C Split-Hinge Tube Furnaces
Lindberg/Blue M™ 1200°C Split-Hinge Tube Furnaces
Actual product may vary
Lindberg/Blue M™ 1200°C Split-Hinge Tube Furnaces
Configure this split-hinge tube furnace for horizontal or vertical use. Features independent digital temperature control modules for ease of use.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeated ZoneLength (Metric)Voltage
HTF55667C914 mm53.3 cm208/240 V
HTF55347C
also known as AVL
610 mm43.2 cm208/240 V
Catalog number HTF55667C
Price (SGD)
18,263.00
EA
Add to cart
Heated Zone:
914 mm
Length (Metric):
53.3 cm
Voltage:
208/240 V
Price (SGD)
18,263.00
EA
Add to cart
Configure Thermo Scientific™ Lindberg/Blue M™ 1200°C Split-Hinge Tube Furnaces for horizontal or vertical use. Features independent digital temperature control modules (sold separately) which are available in standard or programmable options.
Key Features
  • Moldatherm™ LGO™ heating element modules for superior radial and linear temperature uniformity and fast heatup and cooldown
  • Long-life, energy-efficient elements require little or no maintenance
  • Unique cabinet design achieves lower exterior surface temperature
  • Heat-reflecting element support assembly creates two highly effective insulating air spaces
  • Compact cabinet with high temperature-resistant painted finish
  • Accepts interchangeable Moldatherm tube adapters
  • Long-life Platinel™ II thermocouple(s) with 10ft. compensated lead wire and polarized plug
Three Zone Models
  • Three independent power circuits (zones) with independent thermocouples for control references
  • Full adjustment of each zone over entire operating range to 1200°C
  • Center zone uniformity achieved and operating length maximized through adjustable profiling of end zones by independent controller
  • Temperature uniformity achieved with independent setpoint of end zones higher or lower than center
Specifications
Heated Zone914 mm
Temperature (Metric)100°C to 1200°C
TypeMultisegment
DisplayLED
Temperature ControlIndependent CC58434C
Dimensions (L x W x H) Exterior21 x 49 x 20 in. (53.3 x 124.5 x 50.8 cm)
Length (English)21 in.
Length (Metric)53.3 cm
Width (English)49 in.
Width (Metric)124.5 cm
Height (English)20 in.
Height (Metric)50.8 cm
Diameter (English) Process Tube3 to 6 in.
Diameter (Metric) Process Tube76.2 to 152.4 cm
Shipping Weight (English)310 lb.
Shipping Weight (Metric)141 kg
Electrical Requirements208/240 V 50/60 Hz
Voltage208/240 V