Isotemp™ Hot Plate, Ambient to 400°C, Ceramic
Isotemp™  Hot Plate, Ambient to 400°C, Ceramic
Actual product may vary
Isotemp™ Hot Plate, Ambient to 400°C, Ceramic
Shape: Square; Size: 10.25 x 10.25 in.; Plug Type: UK/AUS/CN-Style
Have Questions?
Catalog NumberTop Plate Material
HP88850206Ceramic
Catalog number HP88850206
Price (SGD)
-
Top Plate Material:
Ceramic
Experience exceptional safety and temperature performance for your routine protocols. Available in three sizes, providing flexibility from microscale chemistries to production operations.
Advanced technology
  • Strong magnetic coupling
  • Temperature adjustable in 1 degree increments
  • HOT TOP warning system designed to protect from accidental burns with prominent display when heating surface is above 50°C (122°F)
  • Automatic over-temperature protection
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional temperature probe and splash guard protection shield
  • Variable sizes for different sample volumes
Specifications
DescriptionHotplate
Top Plate MaterialCeramic
Size10.25 x 10.25 in.
Temperature (Metric)Ambient to 400°C
Height (English)4 in.
Length (English)16.2 in.
Width (English)11.3 in.
Height (Metric)10.2 cm
Length (Metric)41.1 cm
Width (Metric)28.7 cm
Electrical Requirements230 V, 50/60 Hz
Frequency50/60 Hz
Plug TypeUK-Style, AUS-Style, CN-Style
ShapeSquare
Voltage230 V
TypeHot Plate
Unit SizeEA