Explosion-Proof SAFE-T HP6 Hot Plate, 22°C to 38°C, Aluminum
Explosion-Proof SAFE-T HP6  Hot Plate, 22°C to 38°C, Aluminum
Actual product may vary
Explosion-Proof SAFE-T HP6 Hot Plate, 22°C to 38°C, Aluminum
Shape: Square; Size: 6.13 x 6.13 in.; Max. Sample Load: 11.3 kg
Have Questions?
Catalog NumberVoltageFrequencyWattage
HP11515B120 V50/60 Hz600 w
Catalog number HP11515B
Price (SGD)
2,730.00
EA
Add to cart
Voltage:
120 V
Frequency:
50/60 Hz
Wattage:
600 w
Price (SGD)
2,730.00
EA
Add to cart
Perform applications in Class I/Group D atmospheres with Thermo Scientific™ Explosion-Proof SAFE-T HP6 Hotplates. These hotplates feature overtemperature safety monitors, precise thermostatic controls and sealed aluminum housing for hazardous applications.
Features:
  • Thermostatic safety is set at 243°C (469.4°F) for added protection
  • Maintains temperature within ±5.5°C (9.9°F)
  • Cast aluminum top plate maximizes heat transfer and uniformity (±6.5°C or 11.7°F)
  • Corrosion-resistant steel case for easy cleaning and maintenance
  • Accommodates up to 25 lb. (11.3kg) loads and metal vessels/containers
  • Time to boil: 600mL water in 60 minutes
Recommended for:
  • Recrystallizations utilizing organic solvents
  • Dry/Heat-treat silicon wafers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description120V 50/60Hz; 600w; 5A
Top Plate MaterialAluminum
Size6.13 x 6.13 in.
Temperature (Metric)38°C to 220°C
Wattage600 w
Amperage5 A
Electrical Requirements120 V, 50/60 Hz
Frequency50/60 Hz
Max. Sample Load11.3 kg
ShapeSquare
Voltage120 V
TypeHot Plate
Certifications/ComplianceCOA
Dimensions (L x W x H)6.13 in. x 6.13 in.
Unit SizeEA