Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Actual product may vary
Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Accessories for Thermo Scientific™ Refrigerated and Heated Bath Circulators
Enhance the operation of Refrigerated and Heated Bath Circulators with accessories that include bridges, platforms, cooling coils and connectivity solutions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
HAU160-0078Stainless-steel bridge to hold SC controller on the W13, W15, W26, W45, W46 baths
1600027RS232 Adapter
HAU160-0027RS232 Adapter
Catalog number HAU160-0078
Price (SGD)
-
Description:
Stainless-steel bridge to hold SC controller on the W13, W15, W26, W45, W46 baths
Enhance the operation of Refrigerated and Heated Bath Circulators with Thermo Scientific™ Accessories which include bridges, platforms, cooling coils and connectivity solutions.
  • Warranty varies by country, please contact us for details
Specifications
DescriptionStainless-steel bridge to hold SC controller on the W13, W15, W26, W45, W46 baths
For Use With (Equipment)Thermo Scientific™ Refrigerated and Heated Bath Circulators
TypeBridge
Unit SizeEach