4-Carboxy-3-fluorobenzeneboronic acid, 98%
4-Carboxy-3-fluorobenzeneboronic acid, 98%
4-Carboxy-3-fluorobenzeneboronic acid, 98%
4-Carboxy-3-fluorobenzeneboronic acid, 98%
4-Carboxy-3-fluorobenzeneboronic acid, 98%
CAS: 120153-08-4 | C7H6BFO4 | 183.929 g/mol
Have Questions?
Catalog NumberQuantity
H53285.065 g
Catalog number H53285.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Carboxy-3-fluorobenzeneboronic acid
CAS120153-08-4
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
4-Carboxy-3-fluorobenzeneboronic acid is used as intermediates. Also used in the coupling reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Carboxy-3-fluorobenzeneboronic acid is used as intermediates. Also used in the coupling reaction.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Green, I. R.; et al. 2-Arylnaphthoquinone analogues: potential anti-TB and pro-apoptotic agents. Journal name. 2011, 192-212.