Owl™ Gel Staining Box
Actual product may vary
Owl™ Gel Staining Box
Stain, fix and de-stain in the same container with Thermo Scientific™ Owl™ Gel Staining Boxes, offering a convenient, conventional staining method.
Have Questions?
Catalog NumberQuantity
GSB-3Pk. of 5 boxes
Catalog number GSB-3
HSN 39269099
Price (SGD)
99.00
Special offer
Ends: 30-Jun-2026
123.00
Save 24.00 (20%)
Each
Quantity:
Pk. of 5 boxes
Price (SGD)
99.00
Special offer
Ends: 30-Jun-2026
123.00
Save 24.00 (20%)
Each
Stain, fix and de-stain in the same container with Thermo Scientific™ Owl™ Gel Staining Boxes, offering a convenient, conventional staining method. Boxes hold up to 125mL of liquid and one gel when used on a shaking platform.
  • Trays available in convenient size to hold the proper volume of staining reagents
  • Grooved lid enables proper orientation and stability when stacking
  • Made of clear plastic, so gels can be viewed directly in the tray
Ordering Information:
Package of five
Specifications
DescriptionGel Staining Box; Pack of 5
QuantityPk. of 5 boxes
Volume (Metric)125 mL
Length (Metric)11 cm
Width (Metric)11 cm
Height (Metric)3 cm
Unit SizeEach