SUB Aqua Pro Water Bath
Actual product may vary
Grant
SUB Aqua Pro Water Bath
Includes non-drip clear lid for improved performance, reduced evaporation and increased energy conservation. Grant Instruments™ SUB Aqua Pro Water Bath provides clear, wide-angle viewing LED display for instant reassurance of bath status.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Model
GRASAP2S2 LSAP2s
GRASAP1212 LSAP12
GRASAP1818 LSAP18
GRASAP22 LSAP2
GRASAP2626 LSAP26
FKS151470255 LSAP5
GRASAP55 LSAP5
GRASAP3434 LSAP34
GRASAPD5 and 12 LSAPD
Catalog number GRASAP2S
Price (SGD)
2,943.00
Each
Capacity (Metric):
2 L
Model:
SAP2s
Price (SGD)
2,943.00
Each
Includes non-drip clear lid for improved performance, reduced evaporation and increased energy conservation. Grant Instruments™ SUB Aqua Pro Water Bath provides clear, wide-angle viewing LED display for instant reassurance of bath status.
  • Ambient +5 to 99°C operation
  • Set and Forget™ technology - fast heat-up, accurate temperature control
  • Stability ±0.2°C
  • 0.1°C setting resolution
  • Adjustable over temperature alarm - protect samples from over heating
  • Advanced dry start and run dry protection
  • Suitable for use with heat transfer beads (excluding SAP2 & SAP2S models)
  • Drain tap and transparent lid included
  • Timer and presets
  • 120V & 230V models available
  • 3 year warranty
Specifications
DescriptionWater Bath
Capacity (Metric)2 L
Certifications/ComplianceDIN 12876-3
Temperature RangeAmbient +5° to 99°C
For Use With (Application)Sample preparation, QA/QC samples, warming samples
Heating Capacity350 w (230 AND 120V)
IncludesDrain tap, tray and lid
ModelSAP2s
Temperature PresetsThree
Temperature Stability±0.2°C
Warranty3 Years
Width (Metric)335 mm
Width (Metric) Interior289 mm
Height (Metric)260 mm
Depth (Metric)215 mm
Depth (Metric) Interior139 mm
Shipping Weight (Metric)4 kg
Voltage120/230 V
Wattage0.35 kW