Adjustable Angle Platform Rocker
Actual product may vary
Grant
Adjustable Angle Platform Rocker
Large capacity, adjustable angle and speed platform rocker, providing soft or intensive side to side rocking for optimal mixing of samples. Suitable for use in cold rooms or incubators with ambient operating temperature range +4ºC to 40ºC.
Have Questions?
Catalog NumberSpeed Range
GRAPMR-1001 to 99 rpm
Catalog number GRAPMR-100
Price (SGD)
4,852.00
Each
Speed Range:
1 to 99 rpm
Price (SGD)
4,852.00
Each
  • Tilt angle range, adjustable up to 10º
  • Loads up to 5kg
  • Variable speed: 1 to 99 oscillations per minute
  • Continuous or timed operation, with automatic switch-off
  • Smooth, non-slip mat supplied as standard
  • Compact, sturdy construction with a low profile and small footprint
Specifications
Rocking Angle0 to 10°
Load Bearing Capacity (Metric)5 kg
Speed Range1 to 99 rpm
Voltage120/230 V
Temperature Range (Metric)4°C to 40°C
Height (Metric)250 mm
Length (Metric)480 mm
Width (Metric)400 mm
Platform StyleAdjustible Tilt
Timer Range1 min. to 99:59 hr.
Unit SizeEach