JB Nova Digital Water Bath with Lid
Actual product may vary
JB Nova Digital Water Bath with Lid
JB Nova Digital Water Bath with Lid
Grant
JB Nova Digital Water Bath with Lid
Provides stable temperature control, simple interface and fast heat-up. Grant Instruments™ JB Nova Digital Water Bath with Lid is available in a choice of four models with a base tray and lid included as standard.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Dimensions (L x W x H)Heating Capacity
GRAJBN1818 L570 x 335 x 275 mm1400 w (230V) 1050 w (120V)
GRAJBN55 L215 x 335 x 200 mm350 w (120V)
Catalog number GRAJBN18
Price (SGD)
3,415.00
Each
Capacity (Metric):
18 L
Dimensions (L x W x H):
570 x 335 x 275 mm
Heating Capacity:
1400 w (230V) 1050 w (120V)
Price (SGD)
3,415.00
Each
Provides stable temperature control, simple interface and fast heat-up. Grant Instruments™ JB Nova Digital Water Bath with Lid is available in a choice of four models with a base tray and lid included as standard.
  • Temperature range ambient +5°C to 95°C operation
  • Stability: ±0.5°C
  • Unique Set and ForgetTM technology - fast heat-up, reliable temperature control
  • Simple, intuitive controls - quick and easy to set temperature
  • Practical front panel lock - disables front panel controls preventing unintentional changes
  • Suitable for use with heat transfer beads
  • Drain tap on 12L, 18L and 26L baths
  • 3 year warranty
Specifications
Capacity (Metric)18 L
Temperature RangeAmbient +5° to 95°C
Heating Capacity1400 w (230V) 1050 w (120V)
Temperature Resolution0.5°C
Temperature Stability±0.5 at 70°C
Warranty3 Years
Dimensions (L x W x H)570 x 335 x 275 mm