Check Order Status
Trifluoroacetic Acid, for DNA Synthesis And Protein Sequencing, Fisher Chemical™
Element Search
Structure Search
Trifluoroacetic Acid, for DNA Synthesis And Protein Sequencing, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
GEET3255PB05
also known as GEET/3255/PB05
100 mL
Catalog number
GEET3255PB05
also known as GEET/3255/PB05
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
pH
0.45
CAS
76-05-1
Melting Point
-15°C
Boiling Point
72°C
Color
Colorless
View more
Trifluoroacetic Acid, for DNA Synthesis And Protein Sequencing