Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™
Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™
Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™
Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™
Trichloroacetic Acid, Certified AR for Analysis, Fisher Chemical™
C2HCl3O2, CAS Number-76-03-9, trichloressigsaeure, konesta, amchem grass killer, acido tricloroacetico, acide trichloracetique, aceto-caustin, acetic acid, trichloro, trichloroethanoic acid, trichloroacetic acid, trichloroacetic acid, 500g, 196 deg.C, CHEBI:30956, White, 163.39g/mol | CAS: 76-03-9 | C2HCl3O2 | 163.378 g/mol
Have Questions?
Catalog NumberQuantity
GEET300060
also known as GEET/3000/60
1 kg
Catalog number GEET300060
also known as GEET/3000/60
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
pH1.2
CAS76-03-9
Melting Point52°C
Boiling Point196°C
ColorWhite
View more
C2HCl3O2, CAS Number-76-03-9, trichloressigsaeure, konesta, amchem grass killer, acido tricloroacetico, acide trichloracetique, aceto-caustin, acetic acid, trichloro, trichloroethanoic acid, trichloroacetic acid, trichloroacetic acid, 500g, 196 deg.C, CHEBI:30956, White, 163.39g/mol