Check Order Status
Toluene, 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Element Search
Structure Search
Toluene, 99.8+%, for Residue Analysis, Distol™, Fisher Chemical™
Have Questions?
Catalog Number
Quantity
GEET232317
also known as GEET/2323/17
2.5 L
Catalog number
GEET232317
also known as GEET/2323/17
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Request bulk or custom format
Specifications
Melting Point
-95°C
CAS
108-88-3
Color
Colorless
Boiling Point
111°C
Quantity
2.5 L
View more