Toluene, Optima™, Fisher Chemical™
Toluene, Optima™, Fisher Chemical™
Toluene, Optima™, Fisher Chemical™
Toluene, Optima™, Fisher Chemical™
Toluene, Optima™, Fisher Chemical™
High purity and versatile solvent suitable for HPLC, spectrophotometry, and pesticide residue analysis
Have Questions?
Catalog NumberQuantityPackaging
GEET230617
also known as GEET/2306/17
2.5 LAmber Glass Bottle
Catalog number GEET230617
also known as GEET/2306/17
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Amber Glass Bottle
Request bulk or custom format
Specifications
Melting Point-95°C
CAS108-88-3
ColorColorless
Boiling Point111°C
Quantity2.5 L
View more
  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Tested by GC-ECD to ensure minimum halogenated residue levels (≤10 ng/L) to demonstrate suitability for pesticide analysis
  • Also meets ACS specifications
  • Intended for research use only