80% Ethanol, Molecular Biology Grade
80% Ethanol, Molecular Biology Grade
80% Ethanol, Molecular Biology Grade
80% Ethanol, Molecular Biology Grade
Thermo Scientific Chemicals
80% Ethanol, Molecular Biology Grade
Ready made ethanol/water blend prequalified for molecular biology research applications. | CAS: 64-17-5 | C2H6O | 46.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
GEET08204K74 L Poly Bottle
Catalog number GEET08204K7
Price (SGD)
610.00
EA
Add to cart
Quantity:
4 L
Packaging:
Poly Bottle
Request bulk or custom format
Price (SGD)
610.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthyl Alcohol
CAS64-17-5
ColorClear, Colorless Liquid
Health Hazard 1Serious eye damage/eye irritation (category 2), Flammable liquids (category 2)
Health Hazard 2Causes serious eye irritation, Highly flammable liquid and vapor
View more
  • 80% ethanol and water blend
  • DNase-, RNase-, and protease-free
  • Ready made ethanol/water blend
  • Pre-qualified for molecular biology research applications
  • Suitable for the purification and precipitation of nucleic acids (DNA and RNA) and proteins
  • Tested for use with Applied Biosystems™ MagMAX™ kits
For Manufacturing and Laboratory Use. Not for beverage use.