Check Order Status
Sucrose, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Element Search
Structure Search
Sucrose, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
C12H22O11, CAS Number-57-50-1, amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar, 1kg, CHEBI:17992, White, 342.29g/mol, CZMRCDWAGMRECN-UGDNZRGBSA-N | CAS: 57-50-1 | C12H22O11 | 342.30 g/mol
Have Questions?
Catalog Number
Quantity
Packaging
GEES860060
also known as GEES/8600/60
1 kg
Plastic powder jar
Catalog number
GEES860060
also known as GEES/8600/60
Price (SGD)
46.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Price (SGD)
46.00
EA
Add to cart
Specifications
CAS
57-50-1
Melting Point
190°C
Color
White
pH
6.5 to 7.5
Packaging
Plastic powder jar
View more
C12H22O11, CAS Number-57-50-1, amerfand, cane sugar, d-sucrose, rohrzucker, saccharose, saccharum, sucrose, sugar, table sugar, white sugar, 1kg, CHEBI:17992, White, 342.29g/mol, CZMRCDWAGMRECN-UGDNZRGBSA-N