Check Order Status
Sodium chloride, meets analytical specification of Ph.Eur
Structure Search
Sodium chloride, meets analytical specification of Ph.Eur
Have Questions?
Catalog Number
Quantity
Packaging
GEES316160
also known as GEES/3161/60
1 kg
Plastic powder jar
Catalog number
GEES316160
also known as GEES/3161/60
Price (SGD)
29.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Price (SGD)
29.00
EA
Add to cart
Specifications
CAS
7647-14-5
Quantity
1 kg
Molecular Formula
ClNa
MDL Number
MFCD00003477
Synonym
sodium chloride, salt, table salt, halite, saline, rock salt, common salt, sodium chloride nacl, dendritis, purex
View more