Check Order Status
Potassium Iodide, Certified AR for Analysis, Fisher Chemical
Element Search
Structure Search
Potassium Iodide, Certified AR for Analysis, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
GEEP588060
also known as GEEP/5880/60
1 kg
Plastic powder jar
Catalog number
GEEP588060
also known as GEEP/5880/60
Price (SGD)
375.00
EA
Add to cart
Quantity:
1 kg
Packaging:
Plastic powder jar
Request bulk or custom format
Price (SGD)
375.00
EA
Add to cart
Specifications
Boiling Point
1330°C
Melting Point
680°C
CAS
7681-11-0
Color
White
pH
6 to 8
View more