Phenolphthalein Solution 1% In Ethanol, Pure, Indicator Grade, Fisher Chemical™
Actual product may vary
Phenolphthalein Solution 1% In Ethanol, Pure, Indicator Grade, Fisher Chemical™
Phenolphthalein Solution 1% In Ethanol, Pure, Indicator Grade, Fisher Chemical™
Phenolphthalein Solution 1% In Ethanol, Pure, Indicator Grade, Fisher Chemical™
C20H14O4, CAS Number-77-09-8, 500mL, CHEBI:34914, Yellow, KJFMBFZCATUALV-UHFFFAOYSA-N, 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one, 5913, 318.328, Amber glass bottle, Liquid, 4764, C1-CC-C2C(-C1)C(-O)OC2(C3-CC-C(C-C3)O)C4-CC-C(C-C4)O | CAS: 77-09-8 | C20H14O4 | 318.33 g/mol
Have Questions?
Catalog NumberQuantity
GEEP2401L08
also known as GEEP/2401L/08
500 mL
Catalog number GEEP2401L08
also known as GEEP/2401L/08
Price (SGD)
62.00
Each
Quantity:
500 mL
Price (SGD)
62.00
Each
C20H14O4, CAS Number-77-09-8, 500mL, CHEBI:34914, Yellow, KJFMBFZCATUALV-UHFFFAOYSA-N, 3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one, 5913, 318.328, Amber glass bottle, Liquid, 4764, C1-CC-C2C(-C1)C(-O)OC2(C3-CC-C(C-C3)O)C4-CC-C(C-C4)O
Specifications
CAS77-09-8
Molecular FormulaC20H14O4
MDL NumberMFCD00005913
InChI KeyKJFMBFZCATUALV-UHFFFAOYSA-N
SMILESOC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
IUPAC Name3,3-bis(4-hydroxyphenyl)-2-benzofuran-1-one
Molecular Weight (g/mol)318.33
PubChem CID4764
ChEBICHEBI:34914
GradePure
PackagingAmber glass bottle
ColorYellow
Quantity500 mL
Physical FormLiquid
Unit SizeEach