Check Order Status
Paraffin Wax, Pure, Melting Point about 57°C, Pastillated, Fisher Chemical
Element Search
Structure Search
Paraffin Wax, Pure, Melting Point about 57°C, Pastillated, Fisher Chemical
Have Questions?
Catalog Number
Quantity
Packaging
GEEP065090
also known as GEEP/0650/90
450 g
Plastic powder jar
Catalog number
GEEP065090
also known as GEEP/0650/90
Price (SGD)
-
Quantity:
450 g
Packaging:
Plastic powder jar
Request bulk or custom format
Specifications
CAS
8002-74-2
Melting Point
58°C
Color
White
Boiling Point
322°C
Packaging
Plastic powder jar
View more