Check Order Status
Nitric Acid 68-70% d=1.42, Certified AR, for Analysis, Fisher Chemical™
Element Search
Structure Search
Nitric Acid 68-70% d=1.42, Certified AR, for Analysis, Fisher Chemical™
HNO3, CAS Number-7697-37-2, acide nitrique, azotic acid, hydrogen nitrate, salpetersaeure, rfna, nitryl hydroxide, nitrous fumes, nital, acidum nitricum, aqua fortis, 2.5L, 122 deg.C, CHEBI:48107, Colorless Yellow, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -42 deg.C, 11349, 63.012
Have Questions?
Catalog Number
Quantity
Packaging
GEEN2300PB17
also known as GEEN/2300/PB17
2.5 L
HDPE plastic bottle
Catalog number
GEEN2300PB17
also known as GEEN/2300/PB17
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
HDPE plastic bottle
Request bulk or custom format
Specifications
pH
1
Color
Colorless Yellow
CAS
7697-37-2
Melting Point
-42°C
Boiling Point
122°C
View more
HNO3, CAS Number-7697-37-2, acide nitrique, azotic acid, hydrogen nitrate, salpetersaeure, rfna, nitryl hydroxide, nitrous fumes, nital, acidum nitricum, aqua fortis, 2.5L, 122 deg.C, CHEBI:48107, Colorless Yellow, GRYLNZFGIOXLOG-UHFFFAOYSA-N, nitric acid, -42 deg.C, 11349, 63.012