Check Order Status
Methylated Spirit (IDA 99), 99% (v/v), Pure, (Industrial Methylated Spirit, 74 0.P.)
Element Search
Structure Search
Methylated Spirit (IDA 99), 99% (v/v), Pure, (Industrial Methylated Spirit, 74 0.P.)
Have Questions?
Catalog Number
Quantity
Packaging
GEEM445015
also known as GEEM/4450/15
1 L
Amber glass bottle
Catalog number
GEEM445015
also known as GEEM/4450/15
Price (SGD)
46.00
EA
Add to cart
Quantity:
1 L
Packaging:
Amber glass bottle
Request bulk or custom format
Price (SGD)
46.00
EA
Add to cart
Specifications
Melting Point
-114.1°C
Color
Colorless
Boiling Point
78.2°C
Packaging
Amber glass bottle
MDL Number
3568
View more